siRNA Details
| Gene Symbol: Cers5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028015.2 | NP_082291.1 | 3 | 471 | ||
| GenBank mRNA | AK010241.1 | - | 3 | 399 | ||
| AK075694.1 | - | 3 | 391 | |||
| AK145589.1 | - | 3 | 379 | |||
| AK145762.1 | - | 3 | 393 | |||
| AK146220.1 | - | 3 | 393 | |||
| AK146417.1 | - | 3 | 362 | |||
| AK219835.1 | - | 3 | 181 | |||
| AY029533.1 | - | 3 | 391 | |||
| BC043059.1 | - | 418 | 6419704 | |||
| BC046797.1 | - | 3 | 471 | 6403129 | ||

Cers5
Genomic Map