siRNA Details
| Gene Symbol: Ist1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028018.2 | NP_082294.1 | 4 | 407 | ||
| GenBank mRNA | AK010262.1 | - | 4 | 377 | ||
| AK050468.1 | - | 4 | 374 | |||
| AK088192.1 | - | 4 | 362 | |||
| AK122215.1 | - | 2 | 181 | |||
| AK152830.1 | - | 4 | 374 | |||
| AK161899.1 | - | 4 | 363 | |||
| AK207302.1 | - | 2 | 101 | |||
| AK218302.1 | - | 2 | 39 | |||
| BC017605.1 | - | 4 | 373 | 4507347 | ||
| BC039052.1 | - | 4 | 350 | 2631157 | ||

Ist1
Genomic Map