siRNA Details
| Gene Symbol: Smg9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028047.2 | NP_082323.1 | 5 | 833 | ||
| GenBank mRNA | AK005126.1 | - | 5 | 833 | ||
| AK039368.1 | - | 5 | 832 | |||
| AK141576.1 | - | 5 | 824 | |||
| AK161454.1 | - | 5 | 843 | |||
| AK169651.1 | - | 5 | 757 | |||
| BC034211.1 | - | 5 | 748 | 4988361 | ||

Smg9
Genomic Map