siRNA Details
| Gene Symbol: Synpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163032.1 | NP_001156504.1 | 5 | 2105 | ||
| NM_028052.4 | NP_082328.3 | 6 | 1999 | |||
| GenBank mRNA | AK005132.1 | - | 6 | 1920 | ||
| AK032442.1 | - | 5 | 2007 | |||
| AK049661.1 | - | 5 | 1951 | |||
| AK078223.1 | - | 6 | 2000 | |||
| BC026512.1 | - | 6 | 1946 | 4527827 | ||

Synpr
Genomic Map