siRNA Details
| Gene Symbol: Tmem68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028097.3 | NP_082373.1 | 6, 7 | 928 | ||
| GenBank mRNA | AK008493.1 | - | 6, 7 | 962 | ||
| AK048210.1 | - | 6, 7 | 928 | |||
| AK084881.1 | - | 6, 7 | 962 | |||
| AK088600.1 | - | 7, 8 | 1098 | |||
| AK147765.1 | - | 6, 7 | 961 | |||
| AK162563.1 | - | 6, 7 | 961 | |||
| AK165916.1 | - | 6, 7 | 990 | |||
| AK166237.1 | - | 6, 7 | 1014 | |||
| BC016240.1 | - | 6, 7 | 946 | 4506688 | ||

Tmem68
Genomic Map