siRNA Details
Gene Symbol: Jmjd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028101.4 | NP_082377.2 | 8 | 662 | ||
GenBank mRNA | AK011308.1 | - | 8 | 659 | ||
AK148079.1 | - | 8 | 640 | |||
AK155810.1 | - | 8 | 664 | |||
AK172073.1 | - | 8 | 659 | |||
BC010800.1 | - | 8 | 624 | 4235072 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
