siRNA Details
| Gene Symbol: Tpx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141975.1 | NP_001135447.1 | 10 | 1268 | ||
| NM_001141976.1 | NP_001135448.1 | 10 | 1284 | |||
| NM_001141977.1 | NP_001135449.1 | 10 | 1472 | |||
| NM_001141978.1 | NP_001135450.1 | 10 | 1271 | |||
| NM_028109.4 | NP_082385.3 | 10 | 1459 | |||
| GenBank mRNA | AK011311.2 | - | 7 | 827 | ||
| AK153957.1 | - | 10 | 1206 | |||
| AK167030.1 | - | 7 | 908 | |||
| BC060619.1 | - | 10 | 1360 | 6812536 | ||

Tpx2
Genomic Map