siRNA Details
| Gene Symbol: Cep89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028120.2 | NP_082396.1 | 3 | 346 | ||
| GenBank mRNA | AK012067.1 | - | 3 | 347 | ||
| AK029107.1 | - | 3 | 377 | |||
| AK082303.1 | - | 3 | 333 | |||
| BC003981.1 | - | 3 | 404 | 3490246 | ||

Cep89
Genomic Map