siRNA Details
| Gene Symbol: Slc37a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028123.3 | NP_082399.3 | 2, 3 | 255 | ||
| GenBank mRNA | AK076213.1 | - | 2, 3 | 198 | ||
| AK077836.1 | - | 2, 3 | 199 | |||
| AK145242.1 | - | 2, 3 | 224 | |||
| AK167716.1 | - | 2, 3 | 211 | |||
| BC005744.1 | - | 2, 3 | 220 | |||
| BC099673.1 | - | 2, 3 | 192 | 4936456 | ||
| BC100391.1 | - | 4, 5 | 435 | |||
| CJ161523.1 | - | 2, 3 | 256 | |||

Slc37a3
Genomic Map