siRNA Details
Gene Symbol: Zfp157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028130.3 | NP_082406.2 | 5 | 594 | ||
GenBank mRNA | AK028290.1 | - | 5 | 554 | ||
BC059261.1 | - | 5 | 566 | 6409233 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028130.3 | NP_082406.2 | 5 | 594 | ||
GenBank mRNA | AK028290.1 | - | 5 | 554 | ||
BC059261.1 | - | 5 | 566 | 6409233 |