siRNA Details
| Gene Symbol: Atxn7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028139.5 | NP_082415.2 | 3 | 315 | ||
| GenBank mRNA | AK013145.1 | - | 3 | 222 | ||
| BB843098.1 | - | 3 | 238 | |||
| BC017546.1 | - | 2 | 116 | |||
| BC100598.1 | - | 2 | 127 | |||
| BY019350.1 | - | 2 | 239 | |||
| BY203850.1 | - | 3 | 187 | |||

Atxn7l1
Genomic Map