siRNA Details
Gene Symbol: Atxn7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028139.5 | NP_082415.2 | 4 | 1092 | ||
GenBank mRNA | AK013145.1 | - | 4 | 999 | ||
BC017546.1 | - | 3 | 893 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atxn7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028139.5 | NP_082415.2 | 4 | 1092 | ||
GenBank mRNA | AK013145.1 | - | 4 | 999 | ||
BC017546.1 | - | 3 | 893 |