siRNA Details
| Gene Symbol: 1600014C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085385.1 | NP_001078854.1 | 3 | 859 | ||
| NM_028166.3 | NP_082442.1 | 3 | 667 | |||
| GenBank mRNA | AK032135.1 | - | 3 | 636 | ||
| AK032342.1 | - | 3 | 638 | |||
| AK146474.1 | - | 3 | 647 | |||
| BC010476.1 | - | 3 | 623 | 3673971 | ||
| BC019834.1 | - | 3 | 790 | 5123719 | ||
| BC085480.1 | - | 3 | 605 | 6401538 | ||

1600014C10Rik
Genomic Map