siRNA Details
| Gene Symbol: 1600014C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085385.1 | NP_001078854.1 | 3 | 2183 | ||
| NM_028166.3 | NP_082442.1 | 3 | 1991 | |||
| GenBank mRNA | AK032342.1 | - | 3 | 1962 | ||
| AK046058.1 | - | 1 | 702 | |||
| AK146474.1 | - | 3 | 1972 | |||
| BC010476.1 | - | 3 | 1945 | 3673971 | ||
| BC019834.1 | - | 3 | 2112 | 5123719 | ||
| BC085480.1 | - | 3 | 1929 | 6401538 | ||

1600014C10Rik
Genomic Map