siRNA Details
| Gene Symbol: 1700030K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028170.2 | NP_082446.2 | 2 | 745 | ||
| GenBank mRNA | AK006557.1 | - | 1 | 555 | ||
| AK089918.1 | - | 2 | 686 | |||
| AK133815.1 | - | 2 | 749 | |||
| BC008558.1 | - | 2 | 699 | 3592293 | ||

1700030K09Rik
Genomic Map