siRNA Details
| Gene Symbol: Brf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028193.3 | NP_082469.2 | 18 | 2387 | ||
| GenBank mRNA | AK010890.1 | - | 5 | 636 | ||
| BC037224.1 | - | 18 | 2170 | 3257364 | ||
| BC058112.1 | - | 8 | 983 | |||
| BQ573483.1 | - | 1 | 132 | |||

Brf1
Genomic Map