siRNA Details
| Gene Symbol: Cyth4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028195.3 | NP_082471.2 | 13 | 1372 | ||
| GenBank mRNA | AF079972.1 | - | 13 | 1303 | ||
| AK010908.1 | - | 3 | 305 | |||
| AK152306.1 | - | 13 | 1278 | |||
| AK152909.1 | - | 13 | 1278 | |||
| BC018505.1 | - | 12 | 1196 | |||
| BC139056.1 | - | 13 | 1345 | 8862078 | ||

Cyth4
Genomic Map