siRNA Details
| Gene Symbol: Kif1bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028197.2 | NP_082473.2 | 4 | 691 | ||
| GenBank mRNA | AK035659.1 | - | 4 | 679 | ||
| AK129320.1 | - | 4 | 676 | |||
| AK141324.1 | - | 4 | 699 | |||
| AK149629.1 | - | 4 | 628 | |||
| AK158730.1 | - | 5 | 426 | |||
| AK161007.1 | - | 677 | ||||
| AK212287.1 | - | 2 | 88 | |||
| BC046485.1 | - | 4 | 662 | 5350638 | ||
| BC055307.1 | - | 4 | 645 | 6493547 | ||
| BC090974.1 | - | 4 | 645 | 6774777 | ||

Kif1bp
Genomic Map