siRNA Details
| Gene Symbol: Plxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163608.1 | NP_001157080.1 | 14 | 2378 | ||
| NM_028199.3 | NP_082475.3 | 14 | 2357 | |||
| GenBank mRNA | AF378760.1 | - | 14 | 2291 | ||
| AK036144.1 | - | 14 | 2332 | |||
| AK153684.1 | - | 14 | 2344 | |||
| BB129646.2 | - | 1 | 570 | |||

Plxdc1
Genomic Map