siRNA Details
| Gene Symbol: Tmem175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163531.1 | NP_001157003.1 | 10 | 935 | ||
| NM_001163532.1 | NP_001157004.1 | 10 | 938 | |||
| NM_028223.3 | NP_082499.3 | 10 | 1045 | |||
| GenBank mRNA | AK013879.1 | - | 10 | 868 | ||
| AK034579.1 | - | 10 | 914 | |||
| AK082765.1 | - | 10 | 906 | |||
| AK170273.1 | - | 10 | 888 | |||
| BC005542.1 | - | 10 | 853 | 3590497 | ||

Tmem175
Genomic Map