siRNA Details
| Gene Symbol: Rab38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028238.7 | NP_082514.4 | 3 | 876 | ||
| GenBank mRNA | AK009296.1 | - | 3 | 834 | ||
| AK044783.1 | - | 3 | 839 | |||
| AK054253.1 | - | 3 | 877 | |||
| AK165682.1 | - | 3 | 879 | |||
| AY062237.1 | - | 3 | 836 | |||
| BC089578.1 | - | 3 | 836 | 6762755 | ||

Rab38
Genomic Map