siRNA Details
| Gene Symbol: Zfp131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011832.1 | - | 5 | 600 | ||
| AK030398.1 | - | 5 | 799 | |||
| AK047429.1 | - | 4 | 582 | |||
| AK134257.1 | - | 5 | 599 | |||
| AK135723.1 | - | 4 | 583 | |||
| AK162903.1 | - | 4 | 1022 | |||
| AY092766.1 | - | 4 | 691 | |||
| BC048839.1 | - | 2 | 282 | |||
| BC057991.1 | - | 5 | 640 | 5323559 | ||

Zfp131
Genomic Map