siRNA Details
| Gene Symbol: Zfp131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011832.1 | - | 8 | 1866 | ||
| AK030398.1 | - | 8 | 2065 | |||
| AK088022.1 | - | 1 | 1755 | |||
| AK134257.1 | - | 8 | 1865 | |||
| AK135723.1 | - | 6 | 1951 | |||
| AY092766.1 | - | 6 | 2059 | |||
| BC048839.1 | - | 5 | 1548 | |||
| BC057991.1 | - | 7 | 2008 | 5323559 | ||

Zfp131
Genomic Map