siRNA Details
| Gene Symbol: Tmem55a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028264.4 | NP_082540.1 | 6 | 711 | ||
| GenBank mRNA | AK012054.1 | - | 6 | 671 | ||
| AK033845.1 | - | 6 | 674 | |||
| AK049213.1 | - | 6 | 712 | |||
| AX775959.1 | - | 6 | 640 | |||
| BC021435.1 | - | 6 | 656 | 5029766 | ||

Tmem55a
Genomic Map