siRNA Details
| Gene Symbol: Zfp655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028298.3 | NP_082574.1 | 4 | 469 | ||
| GenBank mRNA | AK012336.1 | - | 4 | 472 | ||
| AK220552.1 | - | 4 | 474 | |||
| BC060660.1 | - | 4 | 505 | 30355779 | ||
| BC062945.1 | - | 4 | 469 | 30357186 | ||

Zfp655
Genomic Map