siRNA Details
| Gene Symbol: Tmem135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028343.4 | NP_082619.3 | 11 | 1172 | ||
| GenBank mRNA | AK013269.1 | - | 11 | 1146 | ||
| AK028401.1 | - | 11 | 982 | |||
| AK028846.1 | - | 11 | 1146 | |||
| AK031818.1 | - | 7 | 591 | |||
| AK047200.1 | - | 11 | 1146 | |||
| AK153175.1 | - | 11 | 1096 | |||
| AY040841.1 | - | 11 | 1022 | |||
| BC033279.1 | - | 11 | 1152 | 4165990 | ||
| BC050931.1 | - | 11 | 1081 | 6811187 | ||

Tmem135
Genomic Map