siRNA Details
| Gene Symbol: Rspo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028351.3 | NP_082627.3 | 1, 2 | 637 | ||
| GenBank mRNA | AB055811.1 | - | 1, 2 | 595 | ||
| AK013366.1 | - | 1, 2 | 586 | |||
| AK076308.1 | - | 584 | ||||
| AK133916.1 | - | 1, 2 | 638 | |||
| AY864332.1 | - | 1, 2 | 98 | |||
| BC103794.1 | - | 1, 2 | 571 | 6517974 | ||
| BC145929.1 | - | 1, 2 | 242 | 40131232 | ||
| BC145931.1 | - | 1, 2 | 242 | 40131234 | ||

Rspo3
Genomic Map