siRNA Details
| Gene Symbol: Ttc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028360.2 | NP_082636.3 | 10 | 1184 | ||
| NM_029704.2 | NP_083980.2 | 9 | 1145 | |||
| GenBank mRNA | AK013368.1 | - | 10 | 1137 | ||
| AK034075.1 | - | 10 | 1174 | |||
| BC026866.1 | - | 8 | 828 | |||
| BC037111.1 | - | 9 | 908 | 5370464 | ||

Ttc19
Genomic Map