siRNA Details
| Gene Symbol: Phykpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028398.2 | NP_082674.1 | 13 | 1637 | ||
| GenBank mRNA | AK013489.1 | - | 13 | 1591 | ||
| AK133235.1 | - | 1584 | ||||
| BC024461.1 | - | 13 | 1591 | 4975449 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phykpl| Gene Symbol: Phykpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028398.2 | NP_082674.1 | 13 | 1637 | ||
| GenBank mRNA | AK013489.1 | - | 13 | 1591 | ||
| AK133235.1 | - | 1584 | ||||
| BC024461.1 | - | 13 | 1591 | 4975449 | ||