siRNA Details
| Gene Symbol: Cnih3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160211.1 | NP_001153683.1 | 2 | 902 | ||
| NM_001160212.1 | NP_001153684.1 | 2 | 902 | |||
| NM_028408.3 | NP_082684.1 | 2 | 902 | |||
| GenBank mRNA | AK013789.1 | - | 2 | 899 | ||
| AK033911.1 | - | 2 | 900 | |||
| BC115640.1 | - | 2 | 528 | 40103001 | ||
| BC115641.1 | - | 2 | 528 | 40103002 | ||
| BG800984.1 | - | 2 | 332 | |||

Cnih3
Genomic Map