siRNA Details
| Gene Symbol: Tmem138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013694.1 | - | 4 | 865 | ||
| AK031921.1 | - | 5 | 802 | |||
| AK050471.1 | - | 5 | 868 | |||
| BC058237.1 | - | 5 | 1129 | 6307765 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem138| Gene Symbol: Tmem138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013694.1 | - | 4 | 865 | ||
| AK031921.1 | - | 5 | 802 | |||
| AK050471.1 | - | 5 | 868 | |||
| BC058237.1 | - | 5 | 1129 | 6307765 | ||