siRNA Details
Gene Symbol: Fam101a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028443.2 | NP_082719.1 | 3 | 973 | ||
GenBank mRNA | AK014112.1 | - | 3 | 1002 | ||
BC053444.1 | - | 3 | 973 | 6336076 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam101a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028443.2 | NP_082719.1 | 3 | 973 | ||
GenBank mRNA | AK014112.1 | - | 3 | 1002 | ||
BC053444.1 | - | 3 | 973 | 6336076 |