siRNA Details
| Gene Symbol: Rwdd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025637.3 | NP_079913.2 | 6 | 1011 | ||
| NM_028456.3 | NP_082732.2 | 4 | 795 | |||
| GenBank mRNA | AF439556.1 | - | 4 | 794 | ||
| AK014130.1 | - | 4 | 733 | |||
| AK134288.1 | - | 4 | 786 | |||
| BC037515.1 | - | 2 | 394 | |||
| BC128287.1 | - | 4 | 794 | 40102432 | ||
| BC128288.1 | - | 6 | 1010 | |||
| BC139340.1 | - | 4 | 794 | 8862362 | ||
| BC139342.1 | - | 4 | 794 | 8862364 | ||

Rwdd3
Genomic Map