siRNA Details
| Gene Symbol: 1700040L02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028491.1 | NP_082767.1 | 6 | 688 | ||
| GenBank mRNA | AK006661.1 | - | 6 | 689 | ||
| AK029622.1 | - | 6 | 688 | |||
| BC087900.1 | - | 6 | 678 | 6774064 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700040L02Rik| Gene Symbol: 1700040L02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028491.1 | NP_082767.1 | 6 | 688 | ||
| GenBank mRNA | AK006661.1 | - | 6 | 689 | ||
| AK029622.1 | - | 6 | 688 | |||
| BC087900.1 | - | 6 | 678 | 6774064 | ||