siRNA Details
| Gene Symbol: Gm10731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077188.1 | - | 1 | 254 | ||
| BY081654.1 | - | 257 | ||||
| Gene Symbol: 1700034I23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006596.1 | - | 1 | 303 | ||
| AK077003.1 | - | 1 | 261 | |||
| BC117739.1 | - | 1 | 269 | 40090015 | ||

Gm10731
Genomic Map