siRNA Details
| Gene Symbol: Ube2d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276397.1 | NP_001263326.1 | 1 | 850 | ||
| GenBank mRNA | AK005969.1 | - | 1 | 760 | ||
| AK133385.1 | - | 1 | 742 | |||
| BC050749.1 | - | 1 | 759 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2d2b| Gene Symbol: Ube2d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276397.1 | NP_001263326.1 | 1 | 850 | ||
| GenBank mRNA | AK005969.1 | - | 1 | 760 | ||
| AK133385.1 | - | 1 | 742 | |||
| BC050749.1 | - | 1 | 759 | |||