siRNA Details
Gene Symbol: Ube2d2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276397.1 | NP_001263326.1 | 1 | 1230 | ||
GenBank mRNA | AK133385.1 | - | 1 | 1122 | ||
BC050749.1 | - | 1 | 1139 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ube2d2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276397.1 | NP_001263326.1 | 1 | 1230 | ||
GenBank mRNA | AK133385.1 | - | 1 | 1122 | ||
BC050749.1 | - | 1 | 1139 |