siRNA Details
| Gene Symbol: Ms4a6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028595.4 | NP_082871.2 | 7 | 898 | ||
| GenBank mRNA | AF237910.1 | - | 7 | 856 | ||
| AK008652.1 | - | 7 | 872 | |||
| AK150397.1 | - | 7 | 795 | |||
| AK152258.1 | - | 7 | 795 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ms4a6c| Gene Symbol: Ms4a6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028595.4 | NP_082871.2 | 7 | 898 | ||
| GenBank mRNA | AF237910.1 | - | 7 | 856 | ||
| AK008652.1 | - | 7 | 872 | |||
| AK150397.1 | - | 7 | 795 | |||
| AK152258.1 | - | 7 | 795 | |||