siRNA Details
Gene Symbol: Ppp1r21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028658.4 | NP_082934.3 | 22 | 2629 | ||
GenBank mRNA | AK003781.1 | - | 5 | 609 | ||
AK028185.1 | - | 22 | 2493 | |||
AK155094.1 | - | 22 | 2461 | |||
AK170254.1 | - | 22 | 2517 | |||
BC106118.1 | - | 6 | 563 | |||
BC137673.1 | - | 22 | 2569 | 8860692 |
