siRNA Details
| Gene Symbol: Fam122c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028671.2 | NP_082947.1 | 3, 4 | 342 | ||
| GenBank mRNA | AK015288.1 | - | 3, 4 | 342 | ||
| AK015916.1 | - | 3, 4 | 352 | |||
| BC147640.1 | - | 3, 4 | 330 | 9056630 | ||
| BC147646.1 | - | 3, 4 | 330 | 9056636 | ||
| BC172114.1 | - | 3, 4 | 330 | 9054400 | ||

Fam122c
Genomic Map