siRNA Details
| Gene Symbol: Ift57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028680.3 | NP_082956.2 | 2 | 364 | ||
| GenBank mRNA | AF274590.1 | - | 2 | 382 | ||
| AK014731.1 | - | 2 | 416 | |||
| AK047217.1 | - | 2 | 364 | |||
| BC138143.1 | - | 2 | 338 | 8861162 | ||
| BC138144.1 | - | 2 | 338 | 8861163 | ||
| DQ266091.1 | - | 2 | 405 | |||

Ift57
Genomic Map