siRNA Details
| Gene Symbol: 1700029M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006510.2 | - | 3 | 915 | ||
| AK015107.1 | - | 2 | 738 | |||
| AK016438.1 | - | 3 | 1032 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_028688.1 | - | 738 | Chr. - - [-] on Build | ||

1700029M20Rik
Genomic Map