siRNA Details
| Gene Symbol: Herc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028705.3 | NP_082981.3 | 2 | 279 | ||
| GenBank mRNA | AK017531.1 | - | 2 | 252 | ||
| BC042574.1 | - | 2 | 291 | 4503584 | ||
| BC145978.1 | - | 2 | 281 | 40131281 | ||
| BC145980.1 | - | 2 | 281 | 40131283 | ||

Herc3
Genomic Map