siRNA Details
| Gene Symbol: Arsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028710.3 | NP_082986.3 | 5 | 1171 | ||
| GenBank mRNA | AK018132.2 | - | 2 | 588 | ||
| AK158726.1 | - | 5 | 1172 | |||
| AK173082.1 | - | 2 | 270 | |||
| BC084731.1 | - | 2 | 861 | 6813261 | ||

Arsg
Genomic Map