siRNA Details
| Gene Symbol: Arsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166177.1 | NP_001159649.1 | 5 | 1031 | ||
| NM_028710.3 | NP_082986.3 | 14 | 2758 | |||
| GenBank mRNA | AK018132.2 | - | 11 | 2175 | ||
| AK158726.1 | - | 14 | 2759 | |||
| AK173082.1 | - | 11 | 1857 | |||
| BC022158.1 | - | 10 | 1588 | |||
| BC039629.1 | - | 5 | 988 | |||
| BC084731.1 | - | 11 | 2448 | 6813261 | ||

Arsg
Genomic Map