siRNA Details
| Gene Symbol: Als2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159948.2 | NP_001153420.2 | 4 | 746 | ||
| NM_028717.6 | NP_082993.4 | 4 | 577 | |||
| NM_146109.3 | NP_666221.1 | 4 | 577 | |||
| GenBank mRNA | AB053307.1 | - | 4 | 565 | ||
| AK029858.1 | - | 4 | 747 | |||
| BC031479.1 | - | 4 | 554 | 3257574 | ||
| BC046828.1 | - | 4 | 538 | 6409924 | ||

Als2
Genomic Map