siRNA Details
| Gene Symbol: Slc35a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028756.4 | NP_083032.2 | 7 | 1622 | ||
| GenBank mRNA | AK003124.1 | - | 7 | 1550 | ||
| AK030566.1 | - | 7 | 1547 | |||
| AK033921.1 | - | 7 | 1589 | |||
| AK085567.1 | - | 7 | 1561 | |||
| AK150825.1 | - | 7 | 1613 | |||
| AK151484.1 | - | 7 | 1613 | |||
| AK170026.1 | - | 7 | 1588 | |||
| BC011115.1 | - | 7 | 1551 | 4013987 | ||

Slc35a5
Genomic Map