siRNA Details
| Gene Symbol: Cep55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164362.1 | NP_001157834.1 | 4 | 723 | ||
| NM_028293.1 | NP_082569.1 | 3 | 347 | |||
| NM_028760.2 | NP_083036.2 | 4 | 699 | |||
| GenBank mRNA | AK004655.1 | - | 4 | 725 | ||
| AK012311.1 | - | 4 | 496 | |||
| AK028216.1 | - | 3 | 333 | |||
| AK088548.1 | - | 4 | 406 | |||
| BC026966.1 | - | 1 | 55 | |||
| BC031396.1 | - | 4 | 652 | 4950530 | ||
| CJ074252.1 | - | 3 | 350 | |||

Cep55
Genomic Map