siRNA Details
| Gene Symbol: Nuak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195025.1 | NP_001181954.1 | 8 | 2301 | ||
| NM_028778.4 | NP_083054.1 | 7 | 2277 | |||
| GenBank mRNA | AK004737.2 | - | 7 | 2265 | ||
| AK033672.1 | - | 7 | 2264 | |||
| AK034082.1 | - | 8 | 2293 | |||
| BC033302.1 | - | 7 | 2251 | 3663595 | ||
| BC046833.1 | - | 8 | 2276 | 6816569 | ||

Nuak2
Genomic Map