siRNA Details
| Gene Symbol: F13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166391.1 | NP_001159863.1 | 4 | 473 | ||
| NM_028784.3 | NP_083060.2 | 4 | 498 | |||
| GenBank mRNA | AK030914.1 | - | 4 | 473 | ||
| AK036403.1 | - | 4 | 499 | |||
| AK049092.1 | - | 4 | 499 | |||
| AK163591.1 | - | 4 | 473 | |||
| AK165311.1 | - | 4 | 499 | |||
| AK170215.1 | - | 4 | 498 | |||
| AK170278.1 | - | 4 | 499 | |||
| AK170860.1 | - | 4 | 499 | |||
| AK171273.1 | - | 4 | 499 | |||
| BC040274.1 | - | 4 | 489 | 3482233 | ||

F13a1
Genomic Map